Learn More
R&L Enterprises™ Wire Gauze with Ceramic Center
Description
- Non-fraying edges
- Durable and long-lasting
- Ceramic center disperses heat evenly
Specifications
Specifications
| Product Type | Wire Gauze |
| Dimensions (L x W) | 125 x 125 mm |
| Includes | Ceramic center |
| Quantity | 10 Pack |
| Length (Metric) | 125 mm |
| Material | Iron |
| Width (Metric) | 125 mm |