Learn More
Manutan Wire rope sling, stainless steel, soft eye, 10mm diameter
Product Code. 12836238
Encompass_Preferred
Product Code. 12836238 Supplier Manutan Supplier No. A341495
Add to basket
Add to basket
  • Manufactured from high grade stainless steel to AISI 316 standards. Thimble eye or soft eye, secured with copper ferrules. Length: 1m. Test certificate supplied.
  • Maximum rope diameter: 10mm