Learn More
Manutan Wire rope strop, galvanised, 6mm diameter, 6x19 soft eye, 2m long
Product Code. 12801549
Encompass_Preferred
Product Code. 12801549 Supplier Manutan Supplier No. A341536
Add to basket
Add to basket
6 x 19 construction wire rope to British Standards. Strops with eye each end. Stamped with serial number and WLL, safety Factor 5:1.
  • WLL (T): 0.4
  • Diameter: 6mm