missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Wiskostatin

Product Code. 18093862
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18093862 50 mg 50mg
18065532 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18093862 Supplier R&D Systems™ Supplier No. 4434/50
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. N-WASP inhibitor; inhibits Arp2/3 activation

Inhibitor of neural Wiskott-Aldrich syndrome protein (N-WASP) activity. Interacts with the regulatory GTPase-binding domain of N-WASP; inhibits activation of Arp2/3 complex by maintaining N-WASP in an inactive conformation. Also inhibits PIP2-induced actin polymerization (EC50 ∽ 4μM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 253449-04-6
Molecular Formula C17H18Br2N2O

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 426.15
Purity >99%
Molecular Weight (g/mol) 426.15
Chemical Name or Material Wiskostatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.