missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Wiskostatin
Description
Inhibitor of neural Wiskott-Aldrich syndrome protein (N-WASP) activity. Interacts with the regulatory GTPase-binding domain of N-WASP; inhibits activation of Arp2/3 complex by maintaining N-WASP in an inactive conformation. Also inhibits PIP2-induced actin polymerization (EC50 ∽ 4μM).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 426.15 |
| Purity | >99% |
| Molecular Weight (g/mol) | 426.15 |
| Chemical Name or Material | Wiskostatin |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?