Learn More
Edu Lab™ Wood Splint
Product Code. 12955941
Change view
Click to view available options
Material:
Wood
Unit Size:
Pack of 1000
Product Code. Material Unit Size
12955941 Wood Pack of 1000
1 options
Product Code. 12955941 Supplier Edu Lab™ Supplier No. VTT12301178
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
For lighting bunsen burners, testing for oxygen or running 'squeaky pop test'.
  • Supplied in bundles of approximately 1000 splints
Specifications
Product Type Splint
For Use With (Equipment) Lighting Bunsen Burners
Material Wood
For Use With (Application) Running the Good Old Squeaky Pop Test, Testing for Oxygen
No. per Pack 1 Ea.