Learn More
Heinz Herenz Wooden Applicator with Single Cotton Bud Single End Tip
Product Code. 30158383
Change view
Click to view available options
Tip Type:
Single End
Unit Size:
Pack of 1000
Product Code. Tip Type Unit Size
30158383 Single End Pack of 1000
1 options
Product Code. 30158383 Supplier Heinz Herenz Supplier No. 1030118
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Wooden applicator with single cotton bud.
We offer a wide range of sterile and nonsterile swabs:
  • Wood, PS, PP, aluminium or paper applicator
  • Each in various lengths available
  • With cotton, viscose, polyester or foam head
  • Diameter of the heads: small, medium or large
  • This guarantees that the right instrument is available for every special application.
  • For example, the cotton-tipped swabs are recommended for use by ENT specialists.
  • Our sterile cotton buds are all individually wrapped.
Please note in the medical field:
  • Only suitable for single use
  • Use by medical personnel only
  • This applicator size: single cotton bud is 5.0 to 5.5 x 12 mm and wooden stick is 150 x 2.2 mm
Specifications
Autoclavable Not Autoclavable
Handle Material Wood
Product Type Cotton Swab
Tip Material Cotton
Tip Type Single End