Learn More
Heinz Herenz Wooden Applicator Sterile Single Cotton Bud
Product Code. 30158377
Change view
Click to view available options
Tip Type:
Single End
Unit Size:
Pack of 100
Product Code. Tip Type Unit Size
30158377 Single End Pack of 100
1 options
Product Code. 30158377 Supplier Heinz Herenz Supplier No. 1030619
  • ONLINE EXCLUSIVE PRICE
    21.26 €
    19.75 € / Pack of 100
    Save 1.51 €
    7% Off
In Stock
Add to basket
Edge
Add to basket
Sterile wooden applicator with single cotton bud. Double packed.
We offer a wide range of sterile and nonsterile swabs:
  • Wood, PS, PP, aluminium or paper applicator
  • Each in various lengths available
  • With cotton, viscose, polyester or foam head
  • Diameter of the heads: small, medium or large
  • This guarantees that the right instrument is available for every special application.
  • For example, the cotton-tipped swabs are recommended for use by ENT specialists.
  • Our sterile cotton buds are all individually wrapped.
Please note in the medical field:
  • Only suitable for single use
  • Use by medical personnel only
Specifications
Autoclavable Not Autoclavable
Sterility Sterile
Handle Material Wood
Product Type Cotton Swab
Tip Material Cotton
Tip Type Single End