Learn More
Techsit Worktech Laboratory ESD High Chair
Product Code. 30306785
Encompass_Preferred
Change view
Click to view available options
Style:
Footrings
High back
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Style Unit Size
30306785 Footrings Each
30306784 High back Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30306785 Supplier Techsit Supplier No. TS1911
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High ESD vinyl Cleanroom chair
  • ESD Cleanroom vinyl high chair with adjustable footring on ESD glides
  • Black ESD/Anti-static Vinyl that is easy to clean
  • Fully adjustable Seat height, back height, and back rake
  • Ergonomically designed and contoured seat and back
  • Cleanroom class 7 suitable
  • BS EN61340-5-1 Conductivity 104 - 106 Ohms
High ESD Vinly Cleanroom Chair
Specifications
Product Type Cleanroom ESD Chair
Color Black
Autoclavable Not Autoclavable
Back Style Ergonomic
Style Footrings
With Armrest No
With Backrest Yes
With Casters No
Footrest Yes
With Lumbar Support Yes
With Tilt No
Material Vinyl Upholstery
Seating Type High Bench Height