Learn More
Techsit Worktech Laboratory ESD High Chair
Product Code. 30306784
Encompass_Preferred
Click to view available options
Style:
Footrings
High back
Unit Size:
Each
Product Code. 30306784
Brand: Techsit TS1911HB
High ESD vinyl Cleanroom chair
  • ESD Cleanroom vinyl high chair with adjustable footring on ESD glides
  • Black ESD/Anti-static Vinyl that is easy to clean
  • Fully adjustable Seat height, back height, and back rake
  • Ergonomically designed and contoured seat and back
  • Cleanroom class 7 suitable
  • BS EN61340-5-1 Conductivity 104 - 106 Ohms
High ESD Vinly Cleanroom Chair
Specifications
Product Type Cleanroom ESD Chair
Color Black
Autoclavable Not Autoclavable
Back Style Ergonomic
Style High back
With Armrest No
With Backrest Yes
With Casters No
Footrest Yes
With Lumbar Support Yes
With Tilt No
Material Vinyl Upholstery
Seating Type High Bench Height