missing translation for 'onlineSavingsMsg'
Learn More

Techsit Worktech Laboratory Vinyl High Chair

Product Code. 30306778
Encompass_Preferred
Click to view available options
Color:
Black
Blue
Style:
Footrings on glides
High back
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30306778

Brand: Techsit TS1311Blue

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High antimicrobial vinyl Lab chair

  • Vinyl
  • Laboratory high chair with adjustable foot ring on glides
  • Antimicrobial Vinyl that is easy to clean
  • Fully adjustable seat height, back height, and back rake
  • Ergonomically designed and contoured seat and back
  • Cleanroom
  • Class 7 suitable

Specifications

Product Type Lab Swivel Chair
Color Blue
Autoclavable Not Autoclavable
Back Style Ergonomic
Style Footrings on glides
With Armrest No
With Backrest Yes
With Casters No
Footrest Yes
With Lumbar Support Yes
With Tilt No
Material Vinyl Upholstery
Seating Type High Bench Height
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.