missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Wortmannin

Product Code. 30001213
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001213 1 mg 1mg
18702564 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001213 Supplier R&D Systems™ Supplier No. 1232/1
  • ONLINE EXCLUSIVE PRICE
    173.93 €
    156.45 € / 1mg
    Save 17.48 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, irreversible inhibitor of PI 3-kinase. Also inhibitor of PLK1

Potent, selective, cell-permeable and irreversible inhibitor of phosphatidylinositol 3-kinase (PI 3-kinase) (IC50 = 2 - 4 nM). Also potently inhibits polo-like kinase 1 (PLK1) (IC50 = 5.8 nM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19545-26-7
Molecular Formula C23H24O8

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 428.44
Purity >98%
Molecular Weight (g/mol) 428.44
Chemical Name or Material Wortmannin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.