missing translation for 'onlineSavingsMsg'
Learn More

Wortmannin, MedChemExpress

Product Code. 16486397
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486397 5 mg 5mg
16413668 10 mg 10mg
16472816 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16486397 Supplier Medchem Express Supplier No. HY10197
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Wortmannin (SL-2052; KY-12420) is a potent, selective and irreversible PI3K inhibitor with an IC50 of 3 nM. Wortmannin also blocks autophagy formation, and potently inhibits Polo-like kinase 1 (PlK1) and Plk3 with IC50s of 5.8 and 48 nM, respectively.

Wortmannin (SL-2052

Specifications

Chemical Name or Material Wortmannin
CAS 19545-26-7
Color White
Purity Grade Notes Research
Molecular Formula C23H24O8
Quantity 5 mg
Synonym SL-2052 KY-12420
Solubility Information DMSO : ≥ 50 mg/mL (116.71 mM)
SMILES O=C1C([C@@](CC2)([H])[C@@]3(C)C2=O)=C([C@H](OC(C)=O)C3)[C@]4(C)C5=C1OC=C5C(O[C@@H]4COC)=O
Molecular Weight (g/mol) 428.43
Formula Weight 428.43
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.