missing translation for 'onlineSavingsMsg'
Learn More

Wortmannin, TRC

Product Code. 30135011
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135011 25 mg 25mg
30040426 100 mg 100mg
30040469 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135011 Supplier Toronto Research Chemicals Supplier No. TRCW49940025MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 19545-26-7
Molecular Formula C23H24O8
Molecular Weight (g/mol) 428.43
Synonym (+)-Wortmannin, KY 12420, [1S-(1α,6bα,9aβ,11α,11bβ)]-11-(Acetyloxy)-1,6b,7,8,9a,10,11,11b-octahydro-1-(methoxymethyl)-9a,11b-dimethyl-3H-Furo[4,3,2-de]indeno[4,5-h]-2-benzopyran-3,6,9-trione, (1S,6bR,9aS,11R,11bR)-11-(Acetyloxy)-1,6b,7,8,9a,10,11,11b-octahydro-1-(methoxymethyl)-9a,11b-dimethyl-3H-furo[4,3,2-de]indeno[4,5-h]-2-benzopyran-3,6,9-trione, (1S,6bR,9aS,11R,11bR)-1-(Methoxymethyl)-9a,11b-dimethyl-3,6,9-trioxo-3,6,6b,7,8,9,9a,10,11,11b-decahydro-1H-furo[4,3,2-de]indeno[4,5-h]isochromen-11-yl Acetate
SMILES COC[C@H]1OC(=O)c2coc3C(=O)C4=C([C@@H](C[C@@]5(C)[C@H]4CCC5=O)OC(=O)C)[C@]1(C)c23

Specifica

Quantity 25 mg
InChI Formula InChI=1S/C23H24O8/c1-10(24)30-13-7-22(2)12(5-6-14(22)25)16-18(13)23(3)15(9-28-4)31-21(27)11-8-29-20(17(11)23)19(16)26/h8,12-13,15H,5-7,9H2,1-4H3/t12-,13+,15+,22-,23-/m0/s1
Formula Weight 428.1471
Percent Purity >95%
Chemical Name or Material Wortmannin
Grade HPLC
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.