missing translation for 'onlineSavingsMsg'
Learn More

Biochrom WPA Biowave 3+ CT

Product Code. 17223074
Click to view available options
Includes:
Spec, Bluetooth Connectivity
Spec, Bluetooth,Printer
Spec, Built-in Serial Printer
Spectrophotometer
Interface:
TS Display (Color), Bluetooth
Touch Screen Display (Color)
Printer Included:
No
Yes
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17223074

Brand: Biochrom 80300759

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The WPA Biowave 3+ CT offers all the capabilities of a research grade Life Sciences UV/Vis spectrophotometer. Expect exceptionally high resolution with the 3 nm bandwidth. The 5 in. color touch screen allows users to distinguish multiple samples by color on graph overlays and to focus on areas of interest.

  • UV/Vis Spectrophotometer with a 3 nm bandwidth
  • 5 in. color touch screen with zoom and track functions makes it easy to distinguish multiple samples by color on graph overlays
  • Features a USB port for data collection
  • Pre-programmed life science methodologies, including nucleic acid quantification (DNA, RNA and oligonucleotides), protein assays, and cell culture density (OD600) measurements.
  • Loaded with a menu of commonly available fluorescent dyes to specifically target nucleic acids and proteins
  • Nucleic acid scans for purity checking
  • Low volume capability for use with microvolume cuvettes
  • Choose to include a built-in serial printer, Bluetooth connectivity, or both
  • Lightweight and stackable for storage
  • Spectrophotometer has a 2 year warranty and the Xenon flash lamp has a 3 year warranty
  • IQ/OQ available on request

Specifications

Beam Center Height 15 mm
Depth (English) 15.3 in.
Depth (Metric) 390 mm
Detector Type Twin CMOS Array
Display Color Touchscreen
Languages English, French, German, Spanish, Italian, Chinese, Japanese
Light Source Xenon Flash
Product Line WPA Biowave 3+ CT
Spectral Bandwidth 3 nm
Type UV-Vis Spectrophotometer
Voltage 120/240 V
Wavelength Range 190 to 1100 nm
For Use With (Application) UV/Vis Spectrophotometry
For Use With (Equipment) Low Volume UV Cuvettes
Height (English) 3.9 in.
Height (Metric) 100 mm
Includes Spec, Bluetooth Connectivity
Interface Touch Screen Display (Color)
Monochromator Flat Grating
Noise ±0.005 peak to peak ±0.002 RMS
Photometric Linearity ±1.3% or ±0.008 A (Whichever is Greater at 546 nm)
Photometric Range -0.3 to 2.5 A, 0.3 to 199 %T
Photometric Repeatability ±0.002 A to 0.5 A (at 546 nm)
Printer Included No
Shape Round
Stray Light <0.5 %T 340 nm
Wavelength Accuracy ±2 nm
Wavelength Repeatability ±1 nm
Weight (English) 6.6 lb.
Weight (Metric) 3 kg
Width (English) 10 in.
Width (Metric) 260 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.