missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ WR 1065
GREENER_CHOICE

Product Code. 18075812
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18075812 50 mg 50mg
18004872 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18075812 Supplier Tocris Bioscience™ Supplier No. 3356/50
Stock Up and Save This Summer
Use promo code 27950
View all promotions
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

p53 activator. Also ROS scavenger

Cell-permeable reactive oxygen species scavenger that activates p53 through a JNK-dependent signaling pathway. Activates p21waf-1 and MDM2, represses transcription of Myc and thymidine kinase, and inhibits DNA topoisomerase IIα. Induces cell cycle arrest and exhibits cytoprotective activity towards normal cells but not cancer cells in vivo. Also exhibits broad spectrum antiviral activity in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14653-77-1
Molecular Formula C5H16Cl2N2S

Specifications

Quantity 50 mg
Formula Weight 207.16
Purity >98%
Molecular Weight (g/mol) 207.16
Chemical Name or Material WR 1065
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.