missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ WS 12

Product Code. 18045702
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18045702 50 mg 50mg
18045102 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18045702 Supplier R&D Systems™ Supplier No. 3040/50
  • ONLINE EXCLUSIVE PRICE
    882.94 €
    795.90 € / 50mg
    Save 87.04 €
    9% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. TRPM8 agonist; cooling agent

Potent TRPM8 agonist that acts as a cooling agent (EC50 = 193 nM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68489-09-8
Molecular Formula C18H27NO2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 289.41
Purity >99%
Molecular Weight (g/mol) 289.419
Chemical Name or Material WS 12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.