missing translation for 'onlineSavingsMsg'
Learn More

WYE-132, MedChemExpress

Product Code. 16449586
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16449586

Brand: Medchem Express HY10044

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WYE-132 (WYE-125132) is a highly potent, ATP-competitive, and specific mTOR kinase inhibitor (IC50: 0.19±0.07 nM; >5,000-fold selective versus PI3Ks). WYE-132 (WYE-125132) inhibits mTORC1 and mTORC2.

WYE-132 (WYE-125132) is a highly potent, ATP-competitive, and specific mTOR kinase inhibitor (IC50: 0.19±0.07 nM

Specifications

Chemical Name or Material WYE-132
CAS 1144068-46-1
Color Gray
Purity Grade Notes Research
Molecular Formula C27H33N7O4
Quantity 50 mg
Synonym WYE-125132
Solubility Information DMSO : 25 mg/mL (48.11 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(C2=NC(N3CC4CCC(C3)O4)=C5C(N(N=C5)C6CCC7(CC6)OCCO7)=N2)C=C1)NC
Molecular Weight (g/mol) 519.6
Formula Weight 519.6
Percent Purity 95.06%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.