missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ WYE 687 dihydrochloride
GREENER_CHOICE

Product Code. 30001497
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001497

Brand: Tocris Bioscience™ 4282/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective mTOR inhibitor

Potent, ATP-competitive inhibitor of mammalian target of rapamycin (mTOR) (IC50 = 7 nM). Displays selectivity for mTOR over PI 3-Kα (∽100-fold) and PI 3-Kγ (∽500-fold). Inhibits phosphorylation of mTORC1 and mTORC2 substrates including S6K, SGK and Akt; blocks VEGF secretion and HIF-1α expression. Exhibits antiproliferative effects in cancer cell lines through G1 cell cycle arrest and selective apoptosis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1062161-90-3
Molecular Formula C28H32N8O3

Specifications

Quantity 10 mg
Formula Weight 601.53
Purity >98%
Molecular Weight (g/mol) 528.617
Chemical Name or Material WYE 687 dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.