missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ WypAll™ L20 Medium Duty Cleaning Paper Wipes

Product Code. 31188849
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color
31188849 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Plastic packaging - suffocation hazard. Please ensure to carry the product in a safe manner for the weight indicated on the packaging. For food contact.
Product Code. 31188849 Supplier Kimberly-Clark Professional™ Supplier No. 7381
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Plastic packaging - suffocation hazard. Please ensure to carry the product in a safe manner for the weight indicated on the packaging. For food contact.

WypAll™ L20 compact wiping paper is designed for versatile cleaning and wiping in production and maintenance environments, perfect for tool and parts cleanup. The embossing technology of these paper wipes delivers a great balance of performance and value.

No matter which surfaces they come into contact with throughout their day, give your employees and visitors greater peace of mind by providing proven cleaning and disinfecting workplace hygiene solutions to help break the chain of germ transmission. WypAll™ Brand Professional Wiping Paper products are designed for the specific environments where they will be used, to deliver on your wiping task, every time. These blue, 2 ply, single use Medium Duty Cleaning Wipes are versatile, small, easy to handle and designed for a variety of wiping tasks. They are perfect for tool and parts clean-up in process and maintenance environments. Our embossing technology delivers a great balance of performance and value.

  • Individual sheet size 25 (L) x 34 cm (W)
  • Each Wypall™ L20 compact wiping roll contains 165 wiping paper sheets
  • Designed for versatile cleaning and wiping in process and maintenance environments, and perfect for tool and parts clean up
  • Embossing technology in the wiping paper gives these cleaning wipes a great balance of performance and value
  • These WypAll™ paper wipers are in a compact roll format for portability and ease of use
  • Designed for use with the WypAll™ wiping paper dispenser (product code 7041)
  • Help break the chain of germ transmission.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Wiping Paper
Length (English) 9.84 in.
Length (Metric) 25 cm
Width (English) 13.38 in.
Width (Metric) 34 cm
Material Paper
Color Blue
Dimensions (L x W) 25 x 34 cm
Packaging Quantity 12 Rolls x 165 Sheets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.