Learn More
Melford X-a-Gal
Product Code. 30018096
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 30018096
Brand: Melford B719000.25
Chromogenic substrate for detection of a-galactosidase activity in yeast Saccharomyces cerevisiae.
Chromogenic substrate for detection of a-galactosidase activity in yeast Saccharomyces cerevisiae. Used in yeast Two-Hybrid System to detect protein-protein interactions within random libraries. The enzyme a-galactosidase enabels yeast to use disaccharide melibose as a carbon source during growth or fermentation. Secretion of this enzyme in response to activation of the GAL4 gene hydrolizes X-a-GAL and causes yeast colonies to develop a blue color. This reporter system allows immediate visible results on selection plates and eliminates necessity of a second screen filter life assay.
Specifications
Quantity 250 mg