missing translation for 'onlineSavingsMsg'
Learn More

Melford X-a-Gal

Product Code. 30018097
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018097 1 g 1g
30018096 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018097 Supplier Melford Supplier No. B719001.0
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chromogenic substrate for detection of a-galactosidase activity in yeast Saccharomyces cerevisiae.

Chromogenic substrate for detection of a-galactosidase activity in yeast Saccharomyces cerevisiae. Used in yeast Two-Hybrid System to detect protein-protein interactions within random libraries. The enzyme a-galactosidase enabels yeast to use disaccharide melibose as a carbon source during growth or fermentation. Secretion of this enzyme in response to activation of the GAL4 gene hydrolizes X-a-GAL and causes yeast colonies to develop a blue color. This reporter system allows immediate visible results on selection plates and eliminates necessity of a second screen filter life assay.

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.