missing translation for 'onlineSavingsMsg'
Learn More

Klever X-Change Plus (Narrow Cut Head)

Product Code. 17769017
Change view
Click to view available options
Color:
Orange
White
Yellow
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17769017 Yellow Pack of 10
17749017 Orange Pack of 10
17759017 White Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17769017 Supplier Klever Supplier No. PLS200XC20Y
  • ONLINE EXCLUSIVE PRICE
    107.57 €
    92.49 € / Pack of 10
    Save 15.08 €
    14% Off
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recessed blade of the X-Change Plus reduces cut injuries and damaged goods.

  • Cuts boxes, film, tape and thousands more applications.
  • Lighter weight, multiple-position handle for control and reach Innovative tape splitter.
  • Adavnced plastic polymers.
  • Highest safety level.
  • Interchangeable, replaceable head.

Specifications

Blade Material Plastic Polymer
For Use With (Application) Cuts Boxes, Film, Tape, and Thousands More Applications
Disposable Reusable
Sterility Non-sterile
Color Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.