missing translation for 'onlineSavingsMsg'
Learn More

Klever X-Change (Wider Cut Head)

Product Code. 17788997
Change view
Click to view available options
Color:
Blue
Orange
Red
Yellow
Unit Size:
Pack of 12
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17788997 Blue Pack of 12
17798997 Orange Pack of 12
17709007 Red Pack of 12
17719007 Yellow Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17788997 Supplier Klever Supplier No. KCJXC30B
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Recessed blade of the Klever X-Change (Wider Cut Head) reduces cut injuries and damaged goods.

  • Cuts boxes, film, tape and thousands more applications.
  • Ergonomic 'soft touch' handle reduces hand fatigue and stress.
  • Innovative tape splitter.
  • Adavnced plastic polymers.
  • Highest safety level.
  • Interchangeable, replaceable head.

Specifications

Product Type Klever X Change Wide Head
For Use With (Application) Cuts Boxes, Film, Tape, and Thousands More Applications
Disposable Reusable
Sterility Non-sterile
Color Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.