missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Klever X-Change (Wider Cut Head with Scraper)
Click to view available options
Color:
Blue
Orange
Red
Yellow
Unit Size:
Pack of 12
Description
- Cuts boxes, film, tape and thousands more applications.
- Ergonomic 'soft touch' handle reduces hand fatigue and stress.
- Innovative tape splitter.
- Adavnced plastic polymers.
- Highest safety level.
- Interchangeable, replaceable head.
Specifications
Specifications
| For Use With (Application) | Cuts Boxes, Film, Tape, and Thousands More Applications |
| Disposable | Reusable |
| Sterility | Non-sterile |
| Includes | Scraper |
| Color | Blue |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction