missing translation for 'onlineSavingsMsg'
Learn More

Klever X-Change (Wider Cut Head with Scraper)

Product Code. 17759007
Change view
Click to view available options
Color:
Blue
Orange
Red
Yellow
Unit Size:
Pack of 12
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17759007 Yellow Pack of 12
17729007 Blue Pack of 12
17739007 Orange Pack of 12
17749007 Red Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17759007 Supplier Klever Supplier No. KCJXC35Y
  • ONLINE EXCLUSIVE PRICE
    167.00 €
    152.63 € / Pack of 12
    Save 14.37 €
    8% Off
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recessed blade of the X-Change (Wider Cut Head with Scraper) reduces cut injuries and damaged goods.

  • Cuts boxes, film, tape and thousands more applications.
  • Ergonomic 'soft touch' handle reduces hand fatigue and stress.
  • Innovative tape splitter.
  • Adavnced plastic polymers.
  • Highest safety level.
  • Interchangeable, replaceable head.

Specifications

For Use With (Application) Cuts Boxes, Film, Tape, and Thousands More Applications
Disposable Reusable
Sterility Non-sterile
Includes Scraper
Color Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.