Learn More
uvex x-fit XL Hearing Protection Earplugs, Orange, SNR 38 dB
Product Code. 31903210 Shop All Uvex Products
Change view
Click to view available options
Pairs per Package:
150
200
250
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pairs per Package unitSize
31903210 250 Each
31903200 150 Each
31903191 200 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31903210 Supplier uvex Supplier No. 2212143
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
Maximum attenuation and maximum comfort.
  • The ergonomically shaped x-fit XL disposable earplugs are highly noise-reducing and are therefore suitable for use in extremely noisy environments.
  • The soft foam earplugs are very comfortable even when worn for long periods.
TRUSTED_SUSTAINABILITY
Specifications
Size X-Large
Pairs per Package 250
Noise Reduction Rating (NRR) 38 dB
Corded Uncorded
Plug Shape Bullet
Product Type Ear Plugs
Disposable Single-use
Metal Detectable No
Dielectric No