Learn More
ABEBA™ X-Light Black Safety Clog Shoes SB
Product Code. 16378321
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16378321 41 1 pair
16318321 35 1 pair
16328321 36 1 pair
16338321 37 1 pair
16348321 38 1 pair
16358321 39 1 pair
16368321 40 1 pair
16388321 42 1 pair
16398321 43 1 pair
16308331 44 1 pair
16318331 45 1 pair
16328331 46 1 pair
16338331 47 1 pair
16348331 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 16378321 Supplier ABEBA™ Supplier No. 7131035/41
Add to basket
Add to basket
ABEBA™ X-Light Black Safety Clog Shoes SB with silver ions lining to reduce odor, replaceable Active Comfort insole, and a fixed heel strap.
  • Lightweight, comfort, hygiene
  • Comply to EN ISO 20345 : 2012, S2 SRC
  • ESD dissipative according to EN 61340 Climate class 1
  • Slip resistant Mono PU outsole SRC
  • Steel toe cap (200 joules)
  • Black smooth leather
  • Silver ions lining
  • Replaceable Active Comfort insole
  • Sanitized treatment
Specifications
Buckle Material Plastic
Certifications/Compliance CE EN ISO 20345:2012 SB, FO, SRC, ESD
Closure Type Heel strap non-foldable, adjustable
Color Black
Liner Material Breathable lining with Silver ions
Material Leather
Size 41
Sole Features Slip and abrasion resistant, shock absorbing, ESD dissipative
Sole Material Polyurethane
Style Clogs
Toe Material Steel
Packaging Quantity 1 Pair
Certifications/Compliance (Europe) CE EN ISO 20345:2012, DIN EN 61340
Resistant To Water