Learn More
ABEBA™ X-Light White Safety Clog Shoes SB
Product Code. 16338291
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16338291 35 1 pair
16348291 36 1 pair
16358291 37 1 pair
16368291 38 1 pair
16378291 39 1 pair
16388291 40 1 pair
16398291 41 1 pair
16308301 42 1 pair
16318301 43 1 pair
16328301 44 1 pair
16338301 45 1 pair
16348301 46 1 pair
16358301 47 1 pair
16368301 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 16338291 Supplier ABEBA™ Supplier No. 7131030/35
Stock Up Your Lab
Use promo code 28007
View all promotions
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
ABEBA™ X-Light White Safety Clog Shoes SB with silver ions lining to reduce odor, replaceable Active Comfort insole, and fixed heel strap.
  • Lightweight, comfort, hygiene
  • Comply to EN ISO 20345 : 2012, S2 SRC
  • ESD dissipative according to EN 61340 Climate class 1
  • Slip resistant Mono PU outsole SRC
  • Steel toe cap (200 joules)
  • White smooth leather
  • Silver ions lining
  • Replaceable Active Comfort insole
  • Sanitized treatment
Specifications
Buckle Material Plastic
Certifications/Compliance CE EN ISO 20345:2012 SB, SRC, ESD
Closure Type Heel strap non-foldable, adjustable
Color White
Liner Material Breathable lining with Silver ions
Material Leather
Size 35
Sole Features Slip and abrasion resistant, shock absorbing, ESD dissipative
Sole Material Polyurethane
Style Clogs
Toe Material Steel
Packaging Quantity 1 Pair
Certifications/Compliance (Europe) CE EN ISO 20345:2012, DIN EN 61340