Learn More
Agilent Technologies X1 J&W GC packed column in Ulti Metal tubing length 3 ft (0.91 M) 1/8 in . OD 2 mm ID MolSiev
Brand:  Agilent Technologies G3591-81075
Encompass_Preferred
X1 J&W GC packed column in Ulti Metal tubing length 3 ft (0.91 M) 1/8 in . OD 2 mm ID MolSiev
Product Code. 30191091
  • 342.90 € / Pack of 1
Estimated Shipment: 16-09-2026