Learn More
Merck Chemicals X10 Vials Iodoacetamide CAS# 144-48-9
Brand: Merck Chemicals A3221-10VL
X10 Vials Iodoacetamide CAS# 144-48-9
This item is not returnable.
View return policy
Description
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Acute toxicity Category 3
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H301-Toxic if swallowed.
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.