Learn More
Macherey Nagel X100 Chromabond Quechers Mix VI (1.2 g) (0.9 g MgSO4; 0.15 g Chromabond Diamino; 0.15 g C18ec)
Brand:  Macherey Nagel 730974.100
X100 Chromabond Quechers Mix VI (1.2 g) (0.9 g MgSO4; 0.15 g Chromabond Diamino; 0.15 g C18ec)
Product Code. 16490961
  • 222.00 € / Pack of 100