Learn More
Fujifilm Corporation™ RX NIF Sheet X-ray Film
Produces high-quality images from all types of radioisotopes.  Fujifilm Corporation™ RX NIF Sheet X-ray Film is a high-speed, high-contrast film that yields results that are sharp and clear.
Brand:  Fujifilm Corporation™ 16189949
Product Code. 12715325
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Blue-tinted polyester base film produces autoradiographs without the graininess and background fogging associated with clear-based film.
  • RX film can be processed manually or in automatic x-ray film processors.
  • Method and length of exposure depends on the type and amount of radioactive label used.
  • Samples labelled with high-energy β-emitters can be exposed directly or with intensifying screen.
  • Low-energy β-emitters are usually exposed directly
  • Supplied in packs of 100 sheets.
  • Quality autoradiographs
  • Blue sensitive film
  • Resistant to scratches, handling artefacts
  • Superior clarity
  • Very low background Especially suitable for use in DNA sequencing.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.