missing translation for 'onlineSavingsMsg'
Learn More

Meditrade™ Nitril™ 3000 Nitrile Examination Gloves

Product Code. 16682217
Change view
Click to view available options
Size:
Medium
X-Large
Color:
White
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Color unitSize
16682217 X-Large White Pack of 100
16693612 Medium White Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16682217 Supplier Meditrade™ Supplier No. 1280XL

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Non-sterile Meditrade™ disposable nitrile examination gloves provide excellent protection for environments with a high risk of contamination. Powder-free and latex-free they are ideal for use in clinical, laboratory, and manufacturing environments.

  • Examination glove, made of nitrile according to DIN EN 455
  • Protective glove according to PSA
  • Skin-friendly: powder-, protein- and latex-free
  • Micro-roughened for better grip
  • Elastic and tear resistant
  • Ambidextrous fit, with rolled edge
  • Suitable for contact with food
Applications:Suitable for handling cleaning agents and disinfection and for other applications in the laboratory, pharmaceutical industry, professional cleaning, catering and food handling in general.

Specifications

Certifications/Compliance AQL 1.5; EN ISO 13485:2016; 93/42/EWG / EEC: Class 1; PPE Category III; Food Compliance acc. to EC Directive 1935/2004 (blue only)
Description Single use disposable white nitrile gloves
Color White
Disposable Single-use
Latex Free Yes
Material Nitrile
Size X-Large
Thickness (Metric) 0.060 mm (Cuff)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.