missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Small Core Pre-printed GHS CLP Labels

Product Code. 30041411
Click to view available options
Display Text:
1 Red Diamond per Sticker
2 Horizontal Red Diamonds per Sticker
2 Stacked Red Diamonds Plus 2 Additional Red Diamonds per Sticker
2 Stacked Red Diamonds Plus 2 Additional Red Diamons per Sticker
2 Stacked Red Diamonds per Sticker
3 Red Diamonds per Sticker
3 Red Diamonds per Sticker in Rotated Pyramidal Shape
3 Stacked Red Diamonds in Pyramid Formation per Sticker
4 Red Diamonds in Square Formation per Sticker
Width (Metric) Tape:
55 mm
55.00 mm
75.00 mm
105.00 mm
Material:
Paper
Polypropylene
Unit Size:
Pack of 200
Pack of 300
Pack of 400
This item is not returnable. View return policy

Product Code. 30041411

Brand: Brady™ 622317

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Supplied with pre-printed red diamonds, ready for you to print the required warning, compliant to the latest regulations.

For you to print the required warning, compliant to the latest regulations.

Specifications

Certifications/Compliance Compliant with Directive 2011/65/EU and 2015/863
Description Pre-printed Desktop Printer Labels for CLP-GHS Hazardous Substances
Adhesive Type Acrylic, Permanent
Coating None
Color Red on White
Finish Matte
Format Die-cut
Material Paper
Printable Sides Front
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
Display Text 2 Stacked Red Diamonds Plus 2 Additional Red Diamons per Sticker
For Use With (Application) Hazard communication identification
For Use With (Equipment) BBP11, BBP12, PR Plus, THT 25 mm Core Printers
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
Model R7950
No. per Pack 400 Labels/Roll
Quantity 400 Stickers
Shape Rectangle
Size 55.00 mm (W) x 85.00 mm (H)
Temperature Range (English) -40°F to +122°F
Temperature Range (Metric) -40°C to +50°C
Thickness (English) 0.0032 in.
Thickness (Metric) 0.081 mm
Dimensions (LxW) 55.00 mm (W) x 85.00 mm (H)
Length (English) 3.35 in.
Length (Metric) 85.00 mm
Width (English) 2.17 in.
Width (Metric) 55.00 mm
Width (English) Printable Area 55.00 mm
Width (Metric) Tape 55.00 mm
Height (English) 3.35 in.
Height (Metric) 85.00 mm
Alternate Part Number THTCLP-S-L4A-7610-0.4-SC
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.