Learn More
Ratiolab X5 Cryobox w/Grid, PC, Red, 132x132x52mm, Grid 10x10
Brand:  Ratiolab 5150013
X5 Cryobox w/Grid, PC, Red, 132x132x52mm, Grid 10x10
Product Code. 17493713
  • ONLINE EXCLUSIVE PRICE
    67.25 €
    60.42 € / Pack of 5
    Save 6.83 €
    10% Off
Only null left
Estimated Shipment: 09-01-2026