missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xanthan Gum, (Practical Grade), MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| CAS | 11138-66-2 |
| Molecular Formula | C8H14Cl2N2O2 |
| Quantity | 100 g |
| Synonym | 2-2,4-diaminophenoxy ethanol dihydrochloride, 2,4-diaminophenoxyethanol dihydrochloride, 2,4-diaminophenoxyethanol hcl, unii-z5y843j6kq, ethanol, 2-2,4-diaminophenoxy-, dihydrochloride, 2,4-diaminophenoxyethanol 2hcl, 2-2,4-diaminophenoxy ethanol hydrochloride, ethanol, 2-2,4-diaminophenoxy-, hydrochloride 1:2, 4-2-hydroxyethoxy-1,3-phenylenediamine dihydrochloride, 2,4-diaminophenoxyethanol hydrochloride |
| InChI Key | VXYWXJXCQSDNHX-UHFFFAOYSA-N |
| SMILES | C1=CC(=C(C=C1N)N)OCCO.Cl.Cl |
| IUPAC Name | 2-(2,4-diaminophenoxy)ethanol;dihydrochloride |
| Molecular Weight (g/mol) | 241.112 |
| PubChem CID | 47932 |
| Grade | Practical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction