missing translation for 'onlineSavingsMsg'
Learn More

Xanthan Gum (Practical Grade), MP Biomedicals

Product Code. 11482781 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482781 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11482781 Supplier MP Biomedicals Supplier No. 0296002190
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High molecular weight polysaccharide used as a thickening and stabilizing agent in laboratory applications.

Specifications

CAS 11138-66-2
Quantity 500 g
Molecular Formula C8H14Cl2N2O2
Synonym 2-2,4-diaminophenoxy ethanol dihydrochloride, 2,4-diaminophenoxyethanol dihydrochloride, 2,4-diaminophenoxyethanol hcl, unii-z5y843j6kq, ethanol, 2-2,4-diaminophenoxy-, dihydrochloride, 2,4-diaminophenoxyethanol 2hcl, 2-2,4-diaminophenoxy ethanol hydrochloride, ethanol, 2-2,4-diaminophenoxy-, hydrochloride 1:2, 4-2-hydroxyethoxy-1,3-phenylenediamine dihydrochloride, 2,4-diaminophenoxyethanol hydrochloride
InChI Key VXYWXJXCQSDNHX-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1N)N)OCCO.Cl.Cl
IUPAC Name 2-(2,4-diaminophenoxy)ethanol;dihydrochloride
Molecular Weight (g/mol) 241.112
PubChem CID 47932
Grade Practical
Chemical Name or Material Xanthan Gum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.