missing translation for 'onlineSavingsMsg'
Learn More

Xanthene-9-carboxylic Acid, TRC

Product Code. 30455498
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455498 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455498 Supplier Toronto Research Chemicals Supplier No. TRCX4999005G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 82-07-5
Molecular Formula C14 H10 O3
Quantity 5 g
Synonym Xanthene-9-carboxylic acid (6CI,7CI,8CI), 9-Carboxyxanthene, 9H-Xanthen-9-carboxylic acid, NSC 66208, Xanthanoic acid, Xanthenecarboxylic acid, Xanthine-9-carboxylic acid
InChI Formula InChI=1S/C14H10O3/c15-14(16)13-9-5-1-3-7-11(9)17-12-8-4-2-6-10(12)13/h1-8,13H,(H,15,16)
SMILES OC(=O)C1c2ccccc2Oc3ccccc13
IUPAC Name 9H-xanthene-9-carboxylic acid
Molecular Weight (g/mol) 226.23
Formula Weight 226.063
Chemical Name or Material Xanthanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.