missing translation for 'onlineSavingsMsg'
Learn More

Xanthine, 98%

Product Code. 10071600 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10071600 5 g Glass bottle 5g
10081600 25 g Glass bottle 25g
10309233 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10071600 Supplier Thermo Scientific Acros Supplier No. 149170050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 69-89-6 | C5H4N4O2 | 152.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-89-6
Molecular Formula C5H4N4O2
Molecular Weight (g/mol) 152.11
MDL Number MFCD00078453
InChI Key LRFVTYWOQMYALW-UHFFFAOYSA-N
Synonym xanthine, 2,6-dihydroxypurine, xanthin, 2,6-dioxopurine, pseudoxanthine, 1h-purine-2,6 3h,7h-dione, isoxanthine, xanthic oxide, 1h-purine-2,6-diol, 9h-purine-2,6-diol
PubChem CID 1188
ChEBI CHEBI:17712
IUPAC Name 3,7-dihydropurine-2,6-dione
SMILES C1=NC2=C(N1)C(=O)NC(=O)N2

Specifications

Melting Point >300.0°C
Color White to Yellow
Loss on Drying 1% max. (105°C, 5 hrs)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 26, 447
Merck Index 15, 10257
Solubility Information Solubility in water: 1g/14.5l (16°C). Other solubilities: soluble in potassium hydroxide and mineral acids,practically insoluble in most common organic,solvents
Spectrophotometry 98 to 102% (lambda max. 278nm,pH 10) molar absorptivity 9300+/-2%,on dry basis
Formula Weight 152.11
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Xanthine
Show More Show Less
compliance-icons
Product Identifier
  • Xanthine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.