missing translation for 'onlineSavingsMsg'
Learn More

Xanthone, TRC

Product Code. 30454861
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454861 1 g 1g
30450359 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30454861 Supplier Toronto Research Chemicals Supplier No. TRCX5001001G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-47-1
Molecular Formula C13 H8 O2
Molecular Weight (g/mol) 196.2
Synonym Xanthen-9-one (8CI), 9-Oxoxanthene, 9-Xanthone, 9H-Xanthene, 9-oxo-, Benzophenone oxide, Dibenzo-γ-pyrone, Diphenylene ketone oxide, Genicide, NSC 14978, Xanthenone, Xanthone
IUPAC Name xanthen-9-one
SMILES O=C1c2ccccc2Oc3ccccc13

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H8O2/c14-13-9-5-1-3-7-11(9)15-12-8-4-2-6-10(12)13/h1-8H
Formula Weight 196.0524
Chemical Name or Material Xanthone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.