missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies XAV939

Product Code. 17180102
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 17180102

Brand: STEMCELL Technologies 72674

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

WNT pathway inhibitor; Inhibits TNKS1 and TNKS2

  • XAV939 is a potent, small molecule inhibitor of tankyrase (TNKS) 1 and 2 (IC50 = 11 and 4 nM, respectively) (Huang et al.) By inhibiting TNKS activity, XAV939 increases the protein levels of the axin-GSK3β complex and promotes the degradation of β-catenin in SW480 cells (Huang et al.), thereby inhibiting WNT pathway downstream actions
  • Induces cardiomyogenesis in mesoderm progenitor cells derived from mouse embryonic stem cells (Wang et al.)
  • In combination with the SMAD inhibitors LDN193189 and SB431542, promotes induction of forebrain fates in human pluripotent stem cell lines (Maroof et al.)
  • Inhibits colony formation of APC-deficient, β-catenin-dependent DLD-1 colorectal cancer cells (Huang et al.)

Specifications

CAS 284028-89-3
Quantity 10 mg
Molecular Formula C14H11F3N2OS
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.