Learn More
DiscoverX XCT790
Product Code. 17322469
Encompass
Click to view available options
:
5 mg
Product Code. 17322469
Brand: DiscoverX 921139
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
XCT790 is a synthetic biomolecule and potent and selective inhibitor of the estrogen-related receptor alpha (ERRα, IC50=~400nM). Inhibits the signaling activity of PPARγ/ERRα and other ERRα activity. Shows no activity for ERRΔ, Erα, or other nuclear receptors.
XCT790 is a synthetic biomolecule and potent and selective inhibitor of the estrogen-related receptor alpha (ERRα). Inhibits the signaling activity of PPARγ/ERRα and other ERRα activity. Shows no activity for ERRΔ, Erα, or other nuclear receptors.
Specifications
Quantity 5 mg