missing translation for 'onlineSavingsMsg'
Learn More

TNCBio™ XerumFree™

Product Code. 17896927
Change view
Click to view available options
Quantity:
100 mL
500 mL
12 x 500 mL
Unit Size:
Each
Pack of 12
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17896927 100 mL Each
17806937 500 mL Each
17886927 12 x 500 mL Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17896927 Supplier TNCBio™ Supplier No. XF2120100
  • ONLINE EXCLUSIVE PRICE
    159.00 €
    140.00 € / Pack of 1
    Save 19.00 €
    11% Off
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Defined and ACF additive for serum-free cell culture.

Defined additive for serum-free cell culture. Animal/human compound free.

Specifications

Color Transparant liquid
Content And Storage 4°C
Form Liquid
Product Type Cell Culture Supplement
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
Osmolality 285 - 325 mOsm/kg
pH 7 to 8
Quantity 100 mL
Shipping Condition Cold Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.