missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ XL 413 Hydrochloride

Description
XL 413 hydrochloride is a potent and selective Cdc7 inhibitor (IC50 = 3.4 nM). Exhibits >12-fold selectivity for Cdc7 over PIM-1 kinase and >30-fold selectivity over pMCM and CK2. Inhibits proliferation of Colo 205 cells in vitro and attenuates tumor growth of Colo 205 xenografts in mice. XL 413 also improves homology-directed repair (HDR) editing in hematopoietic stem cells (HSC).
Specifications
Specifications
| Chemical Name or Material | 8-Chloro-2-(2S)-2-pyrrolidinylbenzofuro[3,2-d]pyrimidin-4(3H)-one hydrochloride |
| CAS | 1169562-71-3 |
| Quantity | 10 mg |
| Solubility | Soluble to 20mM in water |
| Molecular Formula | C14H12ClN3O2.HCl |
| Formula Weight | Observed MW: 326.18 |
| Purity | >99% |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?