missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ XL 413 Hydrochloride
GREENER_CHOICE

Product Code. p-7112886
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18740298 10 mg 10mg
18721078 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18740298 Supplier Tocris Bioscience™ Supplier No. 5493/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective Cdc7 inhibitor

XL 413 hydrochloride is a potent and selective Cdc7 inhibitor (IC50 = 3.4 nM). Exhibits >12-fold selectivity for Cdc7 over PIM-1 kinase and >30-fold selectivity over pMCM and CK2. Inhibits proliferation of Colo 205 cells in vitro and attenuates tumor growth of Colo 205 xenografts in mice. XL 413 also improves homology-directed repair (HDR) editing in hematopoietic stem cells (HSC).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8-Chloro-2-(2S)-2-pyrrolidinylbenzofuro[3,2-d]pyrimidin-4(3H)-one hydrochloride
CAS 1169562-71-3
Quantity 10 mg
Solubility Soluble to 20mM in water
Molecular Formula C14H12ClN3O2.HCl
Formula Weight Observed MW: 326.18
Purity >99%

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.