missing translation for 'onlineSavingsMsg'
Learn More

CELLPATH XMARK INK SHIELD

Product Code. 30226286
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30226286 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30226286 Supplier CELLPATH Supplier No. RCB030000A
  • ONLINE EXCLUSIVE PRICE
    28.44 €
    13.19 € / 250mL
    Save 15.25 €
    53% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

XMark Ink Shield is a quick-drying fixative spray that is applied to a specimen after marking with XMark Tissue Marking Dye and forms a protective layer on the marked surface while allowing the processing reagent to penetrate the tissue.

  • The chemistry of pigments, especially those seen in yellow, orange and red tissue marking dyes manufactured by alternative supplier's can often suffer during tissue processing protocols.
  • Unique to CellPath, Ink Shield has been specifically designed to use with our XMark range and ensures that specimen margins marked with dyes (including the colours mentioned above) are durable and unaffected during tissue processing.
  • For best results, spray the specimen with XMark Ink Shield from 5-15 cm away.
  • Key Benefits:
  • Quick-drying fixative spray
  • Forms a protective layer on the marked surface
  • Can be used with all colours of XMark Tissue Marking Dye
  • Suitable for all specimen types

Specifications

Grade Histological
Product Type Fixative Spray
Quantity 250 mL
Solution Type Fixative
Form Liquid
compliance-icons compliance-icons
Product Identifier
  • XMark Ink Shield
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P370+P378-In case of fire: Use sand, carbon dioxideor powder extinguisher to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Acetone, ethanol , Methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.