missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Xplorer™ BASIC - Multichannel Trade-In Pipette
GREENER_CHOICE

Product Code. 15260765 Shop All Eppendorf Products
Change view
Click to view available options
Number of Channels:
8
12
Color:
Green
Medium Gray
Orange
Yellow
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Number of Channels Color unitSize
15260765 8 Medium Gray Each
15200775 12 Medium Gray Each
15270765 8 Yellow Each
15210775 12 Yellow Each
15280765 8 Orange Each
15220775 12 Orange Each
15290765 8 Green Each
15230775 12 Green Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 15260765 Supplier Eppendorf™ Supplier No. 4861000570
  • ONLINE EXCLUSIVE PRICE
    880.00 €
    860.00 € / Pack of 1
    Save 20.00 €
    2% Off
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Eliminate potential error risks associated with manual pipetting with pipettes

  • Electronic single, 8 and 12 channel pipettes designed to aspirate and dispense liquid volumes from 0.005 to 10 mL
  • Intuitive operating concept and ergonomic design
  • Multifunctional rocker
  • Selection dial
  • Innovative ejector
  • Ergonomic display angle, Convenient handle
  • Powerful rechargeable battery
  • Flexible charging concept
  • Spring-loaded tip cone
  • Adjustment to external conditions (geographic height, liquids with high density or similar factors)
TRUSTED_SUSTAINABILITY

Especificaciones

Compatible Tips epT.I.P.S.
Adjustable Tip Spacing No
Autoclavable Not Autoclavable
Color Medium Gray
Disposable Reusable
For Use With epT.I.P.S. Pipette Tip (10 μL)
Grip Type Ergonomic
Material Plastic
Number of Channels 8
Programmability Step-based
Sterility Non-sterile
Type Multichannel Pipette
Volume (Metric) 0.5 to 10 μL
For Use With (Application) Pipetting and mixing
For Use With (Equipment) Eppendorf Xplorer Pipettes
Quantity 1 Each
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.