missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Xplorer™ BASIC - Multichannel Trade-In Pipette

Product Code. 15290765 Shop All Eppendorf Products
Change view
Click to view available options
Number of Channels:
8
12
Color:
Green
Medium Gray
Orange
Yellow
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Number of Channels Color Unit Size
15290765 8 Green Each
15260765 8 Medium Gray Each
15200775 12 Medium Gray Each
15270765 8 Yellow Each
15210775 12 Yellow Each
15280765 8 Orange Each
15220775 12 Orange Each
15230775 12 Green Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 15290765 Supplier Eppendorf™ Supplier No. 4861000635
  • ONLINE EXCLUSIVE PRICE
    934.65 €
    860.00 € / Pack of 1
    Save 74.65 €
    7% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eliminate potential error risks associated with manual pipetting with pipettes

  • Electronic single, 8 and 12 channel pipettes designed to aspirate and dispense liquid volumes from 0.005 to 10 mL
  • Intuitive operating concept and ergonomic design
  • Multifunctional rocker
  • Selection dial
  • Innovative ejector
  • Ergonomic display angle, Convenient handle
  • Powerful rechargeable battery
  • Flexible charging concept
  • Spring-loaded tip cone
  • Adjustment to external conditions (geographic height, liquids with high density or similar factors)
TRUSTED_SUSTAINABILITY

Specifications

Compatible Tips epT.I.P.S.
Adjustable Tip Spacing No
Autoclavable Not Autoclavable
Color Green
Disposable Reusable
For Use With epT.I.P.S. Pipette Tip (1200 μL)
Grip Type Ergonomic
Material Plastic
Number of Channels 8
Programmability Step-based
Sterility Non-sterile
Type Multichannel Pipette
Volume (Metric) 50 to 1200 μL
For Use With (Application) Pipetting and mixing
For Use With (Equipment) Eppendorf Xplorer Pipettes
Quantity 1 Each
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.