Learn More
Chemsplash Xtreme 50 SMS Antistatic Disposable Coverall T5/6 Blue

Description
Chemsplash Xtreme SMS 50 Anti-Static Disposable Coverall
- Material: 50GSM Breathable SMS Fabric
- Features:
- Anti-Static Protection
- Elasticated Hood, Back and Ankles
- Knitted Cuffs
- Two-Way Zip With Adhesive Zip Flap
- Silicone and Latex Free
- PFAS Free
- Comes Without the Chest Label
The Chemsplash Xtreme SMS 50 Anti-Static Disposable Coverall is made from breathable SMS fabric chemically treated to offer anti-static protection to EN 1149-5:2008 standards. It also meets CAT III Type 5 and 6. With features such as an elasticated hood, ankles, knitted cuffs, and a two-way zip with an adhesive flap, it provides excellent comfort and robust protection against chemical splash and hazardous particles.
Safety Standards: EN13982-1: TYPE 5, EN13034: TYPE 6, EN1149-5: ANTI-STATIC
Specifications
Specifications
| Product Type | Hooded Coveralls |
| Size | Large |
| Gender | Unisex |
| Material | SMS Fabric |
| Color | Blue |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Knit |
| Neck Style | Hooded |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.